missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-(3-Aminopropyl)morpholine 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 153872500

 View more versions of this product

Catalog No. AC153872500


 

Specifications Description & Specifications

Specifications

N-(3-Aminopropyl)morpholine
123-00-2
97.5
1.4760 to 1.4780 (20°C, 589nm)
Clear
250g
98%
144.22
98%
Authentic
100.0
0.5% max.
Liquid
≥97.5%
C7H16N2O
MFCD00006184

Chemical Identifiers

123-00-2
100.0
C7H16N2O
MFCD00006184
≥97.5%
97.5
98%
144.22
N-(3-Aminopropyl)morpholine, 98%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 204-590-2

RTECSNumber : QD7700000

TSCA : TSCA