Please login to your online account to display your discounted pricing

N-(3-Aminopropyl)morpholine 98%, ACROS Organics™

$177.43 - $177.43

Chemical Identifiers

Chemical Name or Material N-(3-Aminopropyl)morpholine
Name Note 98%
CAS 123-00-2
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C7H16N2O
Formula Weight 144.22
MDL Number MFCD00006184
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC153872500

  SDS
thermo fisher scientific
153872500
Each for $177.43
Description & Specifications

Specifications

Physical Form Liquid
Assay ≥97.5%
Formula Weight 144.22
Quantity 250g
Color Clear
EINECS Number 204-590-2
TSCA TSCA
RTECS Number QD7700000
CAS Max % 100.0
Chemical Name or Material N-(3-Aminopropyl)morpholine
Water 0.5% max.
Name Note 98%
MDL Number MFCD00006184
CAS 123-00-2
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Assay Percent Range 98%
Molecular Formula C7H16N2O
Infrared Spectrum Authentic
CAS Min % 97.5
Refractive Index 1.4760 to 1.4780 (20°C, 589nm)
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.