missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-(3-Aminopropyl)imidazole, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 351942500

 View more versions of this product

Catalog No. AC351942500



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

N-(3-Aminopropyl)imidazole
5036-48-6
100.0
Colorless
296.0°C
Authentic
Glass bottle
1.5175 to 1.5195
250 g
1-3-aminopropyl imidazole, n-3-aminopropyl imidazole, 1h-imidazole-1-propanamine, n-3-aminopropyl-imidazole, 3-1h-imidazol-1-yl propan-1-amine, 3-imidazol-1-ylpropylamine, 3-1h-imidazol-1-yl-1-propanamine, 1-3-aminopropyl-imidazole, 3-imidazol-1-yl propylamine, 3-imidazolylpropylamine
KDHWOCLBMVSZPG-UHFFFAOYSA-N
3-imidazol-1-ylpropan-1-amine
78736
98%
98%
97.5
-68.0°C
1.0620g/mL
154°C
97.5% min. (GC)
C6H11N3
MFCD00009819
1.062
Solubility in water: miscible.
NCCCN1C=CN=C1
125.18
125.17
Liquid

Chemical Identifiers

5036-48-6
125.18
KDHWOCLBMVSZPG-UHFFFAOYSA-N
78736
NCCCN1C=CN=C1
C6H11N3
MFCD00009819
1-3-aminopropyl imidazole, n-3-aminopropyl imidazole, 1h-imidazole-1-propanamine, n-3-aminopropyl-imidazole, 3-1h-imidazol-1-yl propan-1-amine, 3-imidazol-1-ylpropylamine, 3-1h-imidazol-1-yl-1-propanamine, 1-3-aminopropyl-imidazole, 3-imidazol-1-yl propylamine, 3-imidazolylpropylamine
3-imidazol-1-ylpropan-1-amine
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful to aquatic life.
Harmful to aquatic life with long lasting effects.

GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED

GHS Signal Word: Danger

EINECSNumber : 225-730-9

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only