Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(3-Aminopropyl)cyclohexylamine, 98%

Chemical Identifiers

Chemical Name or Material N-(3-Aminopropyl)cyclohexylamine
CAS 3312-60-5
Molecular Formula C9H20N2
Formula Weight 156.27
MDL Number MFCD00003838
UN Number UN2735
Beilstein 741954
Synonym N-Cyclohexyl-1,3-diaminopropane; N-Cyclohexyl-1,3-propanediamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1125014

  SDS
alfa aesar™
L1125014
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1125022

  SDS
alfa aesar™
L1125022
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 156.27
MDL Number MFCD00003838
Molecular Formula C9H20N2
CAS 3312-60-5
Assay 98%
UN Number UN2735
Chemical Name or Material N-(3-Aminopropyl)cyclohexylamine
Synonym N-Cyclohexyl-1,3-diaminopropane; N-Cyclohexyl-1,3-propanediamine
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Beilstein 741954
Health Hazard 2 GHS H Statement
H314-H318-H302-H317
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
May cause an allergic skin reaction.
Melting Point −18°C
Flash Point 111°C (232°F)
Health Hazard 1 Danger
Boiling Point 236°C
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
TSCA TSCA
Quantity 25g
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318-H302-H317
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
May cause an allergic skin reaction.