missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(3-Aminopropyl)cyclohexylamine, 98%

Manufacturer:  Alfa Aesar™ L1125022

 View more versions of this product

Catalog No. AAL1125022



Specifications

Specifications

N-(3-Aminopropyl)cyclohexylamine
236°C
100g
C9H20N2
UN2735
n-3-aminopropyl cyclohexylamine, n1-cyclohexylpropane-1,3-diamine, 1,3-propanediamine, n-cyclohexyl, n-cyclohexyl-1,3-propanediamine, cyclohexyl-1,3-propanediamine, 3-aminopropyl cyclohexylamine, n-cyclohexyl-1,3-diaminopropane, unii-f4xob09fkn, n-cyclohexyl-1,3-propylenediamine, n-cyclohexylpropylene 1,3-diamine
C1CCC(CC1)NCCCN
156.273
156.27
111°C (232°F)
-18°C
3312-60-5
MFCD00003838
741954
ITZPOSYADVYECJ-UHFFFAOYSA-N
N'-cyclohexylpropane-1,3-diamine
18718
98%

Chemical Identifiers

3312-60-5
156.273
ITZPOSYADVYECJ-UHFFFAOYSA-N
18718
C1CCC(CC1)NCCCN
C9H20N2
MFCD00003838
n-3-aminopropyl cyclohexylamine, n1-cyclohexylpropane-1,3-diamine, 1,3-propanediamine, n-cyclohexyl, n-cyclohexyl-1,3-propanediamine, cyclohexyl-1,3-propanediamine, 3-aminopropyl cyclohexylamine, n-cyclohexyl-1,3-diaminopropane, unii-f4xob09fkn, n-cyclohexyl-1,3-propylenediamine, n-cyclohexylpropylene 1,3-diamine
N'-cyclohexylpropane-1,3-diamine
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H302-H317
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
May cause an allergic skin reaction.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.

TSCA : TSCA

Documents