missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-(3'-Aminopropyl)-2-pyrrolidinone, 95%, Thermo Scientific™
Chemical Identifiers
CAS | 7663-77-6 |
---|---|
Molecular Formula | C7H14N2O |
Molecular Weight (g/mol) | 142.20 |
MDL Number | MFCD00003201 |
InChI Key | HJORCZCMNWLHMB-UHFFFAOYSA-N |
Synonym | 1-3-aminopropyl pyrrolidin-2-one, n-3'-aminopropyl-2-pyrrolidinone, 2-pyrrolidinone, 1-3-aminopropyl, 1-3-aminopropyl-2-pyrrolidone, n-3-aminopropyl-2-pyrrolidinone, 1-3-aminopropyl-2-pyrrolidinone, n-3-aminopropyl pyrrolidin-2-one, 1-3-amino-propyl-pyrrolidin-2-one, 1-3-aminoprop-1-yl pyrrolidin-2-one, zlchem 943 |
PubChem CID | 82111 |
IUPAC Name | 1-(3-aminopropyl)pyrrolidin-2-one |
SMILES | NCCCN1CCCC1=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC159600050
|
Thermo Scientific Chemicals
159600050 |
5g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
7663-77-6 | |
142.20 | |
HJORCZCMNWLHMB-UHFFFAOYSA-N | |
82111 | |
NCCCN1CCCC1=O |
C7H14N2O | |
MFCD00003201 | |
1-3-aminopropyl pyrrolidin-2-one, n-3'-aminopropyl-2-pyrrolidinone, 2-pyrrolidinone, 1-3-aminopropyl, 1-3-aminopropyl-2-pyrrolidone, n-3-aminopropyl-2-pyrrolidinone, 1-3-aminopropyl-2-pyrrolidinone, n-3-aminopropyl pyrrolidin-2-one, 1-3-amino-propyl-pyrrolidin-2-one, 1-3-aminoprop-1-yl pyrrolidin-2-one, zlchem 943 | |
1-(3-aminopropyl)pyrrolidin-2-one |
Specifications
7663-77-6 | |
100.0 | |
120.0°C to 123.0°C (1.0 mmHg) | |
Authentic | |
Glass bottle | |
1.4970 to 1.4990 | |
5g | |
1-3-aminopropyl pyrrolidin-2-one, n-3'-aminopropyl-2-pyrrolidinone, 2-pyrrolidinone, 1-3-aminopropyl, 1-3-aminopropyl-2-pyrrolidone, n-3-aminopropyl-2-pyrrolidinone, 1-3-aminopropyl-2-pyrrolidinone, n-3-aminopropyl pyrrolidin-2-one, 1-3-amino-propyl-pyrrolidin-2-one, 1-3-aminoprop-1-yl pyrrolidin-2-one, zlchem 943 | |
NCCCN1CCCC1=O | |
142.20 | |
142.2 | |
N-(3'-Aminopropyl)-2-pyrrolidinone |
94.0 | |
1.0100g/mL | |
>112°C | |
1,5-Diazabicyclo(4.3.0.)non-5-ene: Balance (GC) | |
C7H14N2O | |
MFCD00003201 | |
1.01 | |
HJORCZCMNWLHMB-UHFFFAOYSA-N | |
1-(3-aminopropyl)pyrrolidin-2-one | |
82111 | |
95% |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS Signal Word: Danger
EINECSNumber : 231-632-7
RUO â Research Use Only