missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-(3'-Aminopropyl)-2-pyrrolidinone, 95%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 159600500

 View more versions of this product

Catalog No. AC159600500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

N-(3'-Aminopropyl)-2-pyrrolidinone
>112°C
Glass bottle
1.01
Colorless to Brown
7663-77-6
100.0
C7H14N2O
1-3-amino-propyl-pyrrolidin-2-one, 1-3-aminoprop-1-yl pyrrolidin-2-one, 1-3-aminopropyl pyrrolidin-2-one, 1-3-aminopropyl-2-pyrrolidinone, 1-3-aminopropyl-2-pyrrolidone, 2-pyrrolidinone, 1-3-aminopropyl, n-3'-aminopropyl-2-pyrrolidinone, n-3-aminopropyl pyrrolidin-2-one, n-3-aminopropyl-2-pyrrolidinone, zlchem 943
C1CC(=O)N(C1)CCCN
142.2
142.2
95%
1.0100g/mL
Authentic
1.4970 to 1.4990
120.0°C to 123.0°C (1.0 mmHg)
50g
94.0
1,5-Diazabicyclo(4.3.0.)non-5-ene: Balance (GC)
MFCD00003201
HJORCZCMNWLHMB-UHFFFAOYSA-N
1-(3-aminopropyl)pyrrolidin-2-one
82111
Liquid

Chemical Identifiers

7663-77-6
142.2
HJORCZCMNWLHMB-UHFFFAOYSA-N
82111
C1CC(=O)N(C1)CCCN
C7H14N2O
MFCD00003201
1-3-amino-propyl-pyrrolidin-2-one, 1-3-aminoprop-1-yl pyrrolidin-2-one, 1-3-aminopropyl pyrrolidin-2-one, 1-3-aminopropyl-2-pyrrolidinone, 1-3-aminopropyl-2-pyrrolidone, 2-pyrrolidinone, 1-3-aminopropyl, n-3'-aminopropyl-2-pyrrolidinone, n-3-aminopropyl pyrrolidin-2-one, n-3-aminopropyl-2-pyrrolidinone, zlchem 943
1-(3-aminopropyl)pyrrolidin-2-one
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS Signal Word: Danger

EINECSNumber : 231-632-7

SDS Documents