Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(3,5-Dimethylphenyl)thiourea, 99%

Chemical Identifiers

Chemical Name or Material N-(3,5-Dimethylphenyl)thiourea
CAS 97480-60-9
Molecular Formula C9H12N2S
Formula Weight 180.27
MDL Number MFCD00041173
UN Number UN2811
Beilstein 3540063
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1280603

 
alfa aesar™
L1280603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1280606

 
alfa aesar™
L1280606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 180.27
Chemical Name or Material N-(3,5-Dimethylphenyl)thiourea
MDL Number MFCD00041173
CAS 97480-60-9
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301
Toxic if swallowed.
Health Hazard 3 GHS P Statement
P280f-P301+P310-P330-P501a
Wear protective clothing.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Assay 99%
Molecular Formula C9H12N2S
Melting Point 173° to 175°C
UN Number UN2811
Beilstein 3540063
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P280f-P301+P310-P330-P501a
Wear protective clothing.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301
Toxic if swallowed.