missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(3,5-Dimethylphenyl)thiourea, 99%

Manufacturer:  Alfa Aesar™ L1280606

 View more versions of this product

Catalog No. AAL1280606



Specifications

Specifications

N-(3,5-Dimethylphenyl)thiourea
5g
C9H12N2S
UN2811
1-3,5-dimethylphenyl thiourea, 3,5-dimethylphenyl thiourea, 1-3,5-dimethylphenyl-2-thiourea, n-3,5-dimethylphenyl thiourea, 3,5-dimethyl-phenyl-thiourea, n-3,5-dimethylphenyl thiourea, thiourea,n-3,5-dimethylphenyl, 3,5-dimethylphenyl thiourea, 3,5-dimethylphenyl thiourea, amino 3,5-dimethylphenyl amino methane-1-thione
CC1=CC(=CC(=C1)NC(=S)N)C
180.269
180.27
173°C to 175°C
97480-60-9
MFCD00041173
3540063
XOAYHDJRYDSPJZ-UHFFFAOYSA-N
(3,5-dimethylphenyl)thiourea
736215
99%

Chemical Identifiers

97480-60-9
180.269
XOAYHDJRYDSPJZ-UHFFFAOYSA-N
736215
CC1=CC(=CC(=C1)NC(=S)N)C
C9H12N2S
MFCD00041173
1-3,5-dimethylphenyl thiourea, 3,5-dimethylphenyl thiourea, 1-3,5-dimethylphenyl-2-thiourea, n-3,5-dimethylphenyl thiourea, 3,5-dimethyl-phenyl-thiourea, n-3,5-dimethylphenyl thiourea, thiourea,n-3,5-dimethylphenyl, 3,5-dimethylphenyl thiourea, 3,5-dimethylphenyl thiourea, amino 3,5-dimethylphenyl amino methane-1-thione
(3,5-dimethylphenyl)thiourea
Safety and Handling

Safety and Handling

GHS H Statement
H301
Toxic if swallowed.

GHS P Statement
P280f-P301+P310-P330-P501a
Wear protective clothing.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

Documents