missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-(3,5-Dichlorophenyl)thiourea, 98+%, Thermo Scientific™

$47.55 - $165.05

Chemical Identifiers

CAS 107707-33-5
Molecular Formula C7H6Cl2N2S
Molecular Weight (g/mol) 221.099
MDL Number MFCD00041168
InChI Key BNJARXOEVLYKDN-UHFFFAOYSA-N
Synonym 1-3,5-dichlorophenyl thiourea, 1-3,5-dichlorophenyl-2-thiourea, 3,5-dichlorophenyl thiourea, acmc-1bnq2, amino 3,5-dichlorophenyl amino methane-1-thione, maybridge1_007644, n-3,5-dichlorophenyl thiourea, thiourea, 3,5-dichlorophenyl, thiourea;
PubChem CID 2737539
IUPAC Name (3,5-dichlorophenyl)thiourea
SMILES C1=C(C=C(C=C1Cl)Cl)NC(=S)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1655803
View Documents Promotion Details
Thermo Fisher Scientific
A1655803
1g
Each for $47.55
 
AAA1655806
View Documents Promotion Details
Thermo Fisher Scientific
A1655806
5g
Each for $165.05
 
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

107707-33-5
221.099
BNJARXOEVLYKDN-UHFFFAOYSA-N
2737539
C1=C(C=C(C=C1Cl)Cl)NC(=S)N
C7H6Cl2N2S
MFCD00041168
1-3,5-dichlorophenyl thiourea, 1-3,5-dichlorophenyl-2-thiourea, 3,5-dichlorophenyl thiourea, acmc-1bnq2, amino 3,5-dichlorophenyl amino methane-1-thione, maybridge1_007644, n-3,5-dichlorophenyl thiourea, thiourea, 3,5-dichlorophenyl, thiourea;
(3,5-dichlorophenyl)thiourea

Specifications

107707-33-5
C7H6Cl2N2S
UN2811
1g
BNJARXOEVLYKDN-UHFFFAOYSA-N
(3,5-dichlorophenyl)thiourea
2737539
≥98%
175°C to 179°C
MFCD00041168
3257049
1-3,5-dichlorophenyl thiourea, 1-3,5-dichlorophenyl-2-thiourea, 3,5-dichlorophenyl thiourea, acmc-1bnq2, amino 3,5-dichlorophenyl amino methane-1-thione, maybridge1_007644, n-3,5-dichlorophenyl thiourea, thiourea, 3,5-dichlorophenyl, thiourea;
C1=C(C=C(C=C1Cl)Cl)NC(=S)N
221.099
221.11
N-(3,5-Dichlorophenyl)thiourea
Safety and Handling

Safety and Handling

GHS H Statement
H301
Toxic if swallowed.

P264b-P270-P301+P310-P330-P501c

H301

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Product Certifications