missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(3,4-Dimethylphenyl)-4-fluorobenzenesulfonamide, 97%

Manufacturer:  Alfa Aesar™ H5533003

 View more versions of this product

Catalog No. AAH5533003



Description

Description

Applications
N-(3,4-Dimethylphenyl)-4-fluorobenzenesulfonamide is used in agrochemicals, reagents.

Notes
Keep container tightly closed. Store away from oxidizing agents.
Specifications

Specifications

N-(3,4-Dimethylphenyl)-4-fluorobenzenesulfonamide
288154-65-4
MFCD00784407
IJCSKFYHYFBPNB-UHFFFAOYSA-N
N-(3,4-dimethylphenyl)-4-fluorobenzenesulfonamide
295161
97%
1g
C14H14FNO2S
3,4-dimethylphenyl 4-fluorophenyl sulfonyl amine
CC1=C(C=C(C=C1)NS(=O)(=O)C2=CC=C(C=C2)F)C
279.329
279.33

Chemical Identifiers

288154-65-4
279.329
IJCSKFYHYFBPNB-UHFFFAOYSA-N
295161
CC1=C(C=C(C=C1)NS(=O)(=O)C2=CC=C(C=C2)F)C
C14H14FNO2S
MFCD00784407
3,4-dimethylphenyl 4-fluorophenyl sulfonyl amine
N-(3,4-dimethylphenyl)-4-fluorobenzenesulfonamide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents