Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(3,4-Dichlorophenyl)thiourea, 98+%

Chemical Identifiers

Chemical Name or Material N-(3,4-Dichlorophenyl)thiourea
CAS 19250-09-0
Molecular Formula C7H6Cl2N2S
Formula Weight 221.11
MDL Number MFCD00004935
UN Number UN2811
Beilstein 2102334
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0795806

  SDS
alfa aesar™
L0795806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0795814

  SDS
alfa aesar™
L0795814
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 221.11
Chemical Name or Material N-(3,4-Dichlorophenyl)thiourea
Molecular Formula C7H6Cl2N2S
MDL Number MFCD00004935
Assay ≥98%
Melting Point ≈200°C
CAS 19250-09-0
UN Number UN2811
Beilstein 2102334
Quantity 5g
Odor Odorless
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301
Toxic if swallowed.
Health Hazard 3 GHS P Statement
P280f-P309-P310
Wear protective clothing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P280f-P309-P310
Wear protective clothing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H301
Toxic if swallowed.