Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(3,4-Dichlorophenyl)maleamic acid, 97%

$45.81 - $45.81

Chemical Identifiers

Chemical Name or Material N-(3,4-Dichlorophenyl)maleamic acid
CAS 21395-61-9
Molecular Formula C10H7Cl2NO3
Formula Weight 260.07
MDL Number MFCD00020466
Synonym 4-(3,4-Dichloroanilino)-4-oxo-2-butenoic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2559506

  SDS
alfa aesar™
B2559506
5g Each for $45.81
Description & Specifications

Specifications

Formula Weight 260.07
Chemical Name or Material N-(3,4-Dichlorophenyl)maleamic acid
Assay 97%
Molecular Formula C10H7Cl2NO3
Melting Point 205°C (dec.)
MDL Number MFCD00020466
CAS 21395-61-9
Quantity 5g
Synonym 4-(3,4-Dichloroanilino)-4-oxo-2-butenoic acid
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.