missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-[2-(Trifluoromethyl)phenyl]thiourea, 98%, Thermo Scientific™
$200.44 - $200.44
Chemical Identifiers
CAS | 1736-71-6 |
---|---|
Molecular Formula | C8H7F3N2S |
Molecular Weight (g/mol) | 220.213 |
MDL Number | MFCD00041161 |
InChI Key | FVXFFFHGYOYYQX-UHFFFAOYSA-N |
Synonym | 2-trifluoromethyl phenylthiourea, 1-2-trifluoromethyl phenyl-2-thiourea, 2-trifluoromethyl phenyl thiourea, 1-2-trifluoromethyl phenyl thiourea, 2-trifluoromethyl-phenyl-thiourea, thiourea, 2-trifluoromethyl phenyl, thiourea, n-2-trifluoromethyl phenyl |
PubChem CID | 737227 |
IUPAC Name | [2-(trifluoromethyl)phenyl]thiourea |
SMILES | C1=CC=C(C(=C1)C(F)(F)F)NC(=S)N |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL0937706
|
Thermo Scientific Chemicals
L0937706 |
5 g |
Each for $200.44
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
1736-71-6 | |
220.213 | |
FVXFFFHGYOYYQX-UHFFFAOYSA-N | |
737227 | |
C1=CC=C(C(=C1)C(F)(F)F)NC(=S)N |
C8H7F3N2S | |
MFCD00041161 | |
2-trifluoromethyl phenylthiourea, 1-2-trifluoromethyl phenyl-2-thiourea, 2-trifluoromethyl phenyl thiourea, 1-2-trifluoromethyl phenyl thiourea, 2-trifluoromethyl-phenyl-thiourea, thiourea, 2-trifluoromethyl phenyl, thiourea, n-2-trifluoromethyl phenyl | |
[2-(trifluoromethyl)phenyl]thiourea |
Specifications
1736-71-6 | |
Odorless | |
MFCD00041161 | |
UN2811 | |
2-trifluoromethyl phenylthiourea, 1-2-trifluoromethyl phenyl-2-thiourea, 2-trifluoromethyl phenyl thiourea, 1-2-trifluoromethyl phenyl thiourea, 2-trifluoromethyl-phenyl-thiourea, thiourea, 2-trifluoromethyl phenyl, thiourea, n-2-trifluoromethyl phenyl | |
C1=CC=C(C(=C1)C(F)(F)F)NC(=S)N | |
220.213 | |
220.22 | |
N-[2-(Trifluoromethyl)phenyl]thiourea |
163°C to 165°C | |
C8H7F3N2S | |
5 g | |
2695476 | |
FVXFFFHGYOYYQX-UHFFFAOYSA-N | |
[2-(trifluoromethyl)phenyl]thiourea | |
737227 | |
98% |
Safety and Handling
GHS H Statement
H301
Toxic if swallowed.
P264b-P270-P301+P310-P330-P501c
H301
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber : 217-084-1
TSCA : Yes
Recommended Storage : Ambient temperatures