missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-[2-(Trifluoromethyl)phenyl]thiourea, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals L0937706
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
N-[2-(Trifluoromethyl)phenyl]thiourea | |
163°C to 165°C | |
C8H7F3N2S | |
5 g | |
2695476 | |
FVXFFFHGYOYYQX-UHFFFAOYSA-N | |
[2-(trifluoromethyl)phenyl]thiourea | |
737227 | |
98% |
1736-71-6 | |
Odorless | |
MFCD00041161 | |
UN2811 | |
2-trifluoromethyl phenylthiourea, 1-2-trifluoromethyl phenyl-2-thiourea, 2-trifluoromethyl phenyl thiourea, 1-2-trifluoromethyl phenyl thiourea, 2-trifluoromethyl-phenyl-thiourea, thiourea, 2-trifluoromethyl phenyl, thiourea, n-2-trifluoromethyl phenyl | |
C1=CC=C(C(=C1)C(F)(F)F)NC(=S)N | |
220.213 | |
220.22 |
Chemical Identifiers
1736-71-6 | |
220.213 | |
FVXFFFHGYOYYQX-UHFFFAOYSA-N | |
737227 | |
C1=CC=C(C(=C1)C(F)(F)F)NC(=S)N |
C8H7F3N2S | |
MFCD00041161 | |
2-trifluoromethyl phenylthiourea, 1-2-trifluoromethyl phenyl-2-thiourea, 2-trifluoromethyl phenyl thiourea, 1-2-trifluoromethyl phenyl thiourea, 2-trifluoromethyl-phenyl-thiourea, thiourea, 2-trifluoromethyl phenyl, thiourea, n-2-trifluoromethyl phenyl | |
[2-(trifluoromethyl)phenyl]thiourea |
Safety and Handling
GHS H Statement
H301
Toxic if swallowed.
P264b-P270-P301+P310-P330-P501c
H301
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber : 217-084-1
TSCA : Yes
Recommended Storage : Ambient temperatures