Please login to your online account to display your discounted pricing

N-[2-(Piperidinylamino)ethyl]-4-iodobenzamide, Tocris


σ1 selective ligand

Quantity 50mg

Manufacturer: r & d systems  RDS076350

 View more versions of this product

Catalog No. 076350

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 50mg
Formula Weight 358.22
CAS 155054-42-5
Molecular Formula C14H19IN2O
Chemical Name or Material N-[2-(Piperidinylamino)ethyl]-4-iodobenzamide
Assay Percent Range >98%
Recommended Storage Desiccate at +4°C

Specifications

Chemical Name or Material N-[2-(Piperidinylamino)ethyl]-4-iodobenzamide
Assay Percent Range >98%
Molecular Formula C14H19IN2O
Formula Weight 358.22
Accessories

Desiccate at +4°C