Please login to your online account to display your discounted pricing

N-[2-(Piperidinylamino)ethyl]-4-iodobenzamide, Tocris

σ1 selective ligand

$159.50 - $678.85

Chemical Identifiers

Chemical Name or Material N-[2-(Piperidinylamino)ethyl]-4-iodobenzamide
Assay Percent Range >98%
Molecular Formula C14H19IN2O
Formula Weight 358.22
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

076310

 
r & d systems
RDS076310
10mg Each for $159.50

076350

 
r & d systems
RDS076350
50mg Each for $678.85
Description & Specifications

Specifications

Quantity 10mg
Formula Weight 358.22
Chemical Name or Material N-[2-(Piperidinylamino)ethyl]-4-iodobenzamide
Assay Percent Range >98%
CAS 155054-42-5
Molecular Formula C14H19IN2O
Recommended Storage Desiccate at +4°C
Safety and Handling

Desiccate at +4°C