Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(2-Phenylethyl)thiourea, 97%

$35.55 - $122.05

Chemical Identifiers

Chemical Name or Material N-(2-Phenylethyl)thiourea
CAS 6815-00-5
Molecular Formula C9H12N2S
Linear Formula C6H5(CH2)2NHCSNH2
Formula Weight 180.27
MDL Number MFCD00041194
Beilstein 2092343
Synonym N-Phenethylthiourea
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1052003

  SDS
alfa aesar™
L1052003
1g Each for $35.55

AAL1052006

  SDS
alfa aesar™
L1052006
5g Each for $122.05
Description & Specifications

Specifications

Chemical Name or Material N-(2-Phenylethyl)thiourea
Odor Odorless
Melting Point 136° to 140°C
Quantity 1g
CAS 6815-00-5
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302
Harmful if swallowed.
Health Hazard 3 GHS P Statement
P280f
Wear protective clothing.
Assay 97%
Molecular Formula C9H12N2S
Linear Formula C6H5(CH2)2NHCSNH2
Formula Weight 180.27
MDL Number MFCD00041194
Beilstein 2092343
Synonym N-Phenethylthiourea
Safety and Handling

Warning

GHS P Statement
P280f
Wear protective clothing.

GHS H Statement
H302
Harmful if swallowed.