missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(2-Phenylethyl)thiourea, 97%

Manufacturer:  Alfa Aesar™ L1052003

 View more versions of this product

Catalog No. AAL1052003



Specifications

Specifications

N-(2-Phenylethyl)thiourea
136°C to 140°C
6815-00-5
C6H5(CH2)2NHCSNH2
2092343
OFVGPHQYOCKLLM-UHFFFAOYSA-N
2-phenylethylthiourea
735845
97%
Odorless
1g
C9H12N2S
MFCD00041194
phenethylthiourea, 2-phenylethyl thiourea, n-phenethylthiourea, n-2-phenylethyl thiourea, phenethyl-thiourea, 1-phenethylthiourea, n-2-phenylethyl thioformamide, 1-2-phenylethyl thiourea, amino 2-phenylethyl amino methane-1-thione, 3-phenethylthiourea
C1=CC=C(C=C1)CCNC(=S)N
180.269
180.27

Chemical Identifiers

6815-00-5
180.269
OFVGPHQYOCKLLM-UHFFFAOYSA-N
735845
C1=CC=C(C=C1)CCNC(=S)N
C9H12N2S
MFCD00041194
phenethylthiourea, 2-phenylethyl thiourea, n-phenethylthiourea, n-2-phenylethyl thiourea, phenethyl-thiourea, 1-phenethylthiourea, n-2-phenylethyl thioformamide, 1-2-phenylethyl thiourea, amino 2-phenylethyl amino methane-1-thione, 3-phenethylthiourea
2-phenylethylthiourea
Safety and Handling

Safety and Handling

GHS H Statement
H302
Harmful if swallowed.

GHS P Statement
P280f
Wear protective clothing.

Warning

Documents