Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(2-Nitrophenyl)thiourea, 98+%


Quantity 5g

Manufacturer: alfa aesar™  L1215806

 View more versions of this product

Catalog No. AAL1215806

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 197.22
Chemical Name or Material N-(2-Nitrophenyl)thiourea
Health Hazard 2 GHS H Statement
H301
Toxic if swallowed.
Molecular Formula C7H7N3O2S
CAS 51039-84-0
MDL Number MFCD00041237
Health Hazard 3 GHS P Statement
P280f-P309-P310
Wear protective clothing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Assay ≥98%
Quantity 5g
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Health Hazard 1 Danger
Melting Point 136° to 140°C
UN Number UN2811
Beilstein 2972327

Specifications

Chemical Name or Material N-(2-Nitrophenyl)thiourea
CAS 51039-84-0
Molecular Formula C7H7N3O2S
Formula Weight 197.22
MDL Number MFCD00041237
UN Number UN2811
Beilstein 2972327
Accessories

Danger

GHS P Statement
P280f-P309-P310
Wear protective clothing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H301
Toxic if swallowed.

SDS & Safety Documents