Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(2-Nitrophenyl)thiourea, 98+%


Quantity 1g

Manufacturer: alfa aesar™  L1215803

 View more versions of this product

Catalog No. AAL1215803

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 197.22
Chemical Name or Material N-(2-Nitrophenyl)thiourea
Molecular Formula C7H7N3O2S
Assay ≥98%
MDL Number MFCD00041237
CAS 51039-84-0
Beilstein 2972327
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301
Toxic if swallowed.
Health Hazard 3 GHS P Statement
P280f-P309-P310
Wear protective clothing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Melting Point 136° to 140°C
Quantity 1g
UN Number UN2811

Specifications

Chemical Name or Material N-(2-Nitrophenyl)thiourea
CAS 51039-84-0
Molecular Formula C7H7N3O2S
Formula Weight 197.22
MDL Number MFCD00041237
UN Number UN2811
Beilstein 2972327
Accessories

Danger

GHS P Statement
P280f-P309-P310
Wear protective clothing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H301
Toxic if swallowed.