Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(2-Methoxyphenyl)thiourea, 94%

Chemical Identifiers

Chemical Name or Material N-(2-Methoxyphenyl)thiourea
CAS 1516-37-6
Molecular Formula C8H10N2OS
Formula Weight 182.25
MDL Number MFCD00022162
UN Number UN2811
Beilstein 778949
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1184006

  SDS
alfa aesar™
L1184006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 182.25
CAS 1516-37-6
MDL Number MFCD00022162
Molecular Formula C8H10N2OS
Chemical Name or Material N-(2-Methoxyphenyl)thiourea
Health Hazard 3 GHS P Statement
P280f-P309-P310
Wear protective clothing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Health Hazard 1 Danger
Melting Point 152° to 156°C
Health Hazard 2 GHS H Statement
H301
Toxic if swallowed.
Assay 94%
UN Number UN2811
Quantity 5g
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Odor Odorless
RTECS Number YT6475000
Beilstein 778949
Safety and Handling

Danger

GHS P Statement
P280f-P309-P310
Wear protective clothing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H301
Toxic if swallowed.