Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(2-Methoxyphenyl)maleamic acid, 97%

$32.64 - $366.35

Chemical Identifiers

Chemical Name or Material N-(2-Methoxyphenyl)maleamic acid
CAS 36847-94-6
Molecular Formula C11H11NO4
Formula Weight 221.21
MDL Number MFCD00020460
Synonym 4-(2-Methoxyanilino)-4-oxobut-2-enoic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2475906

 
alfa aesar™
B2475906
5g Each for $32.64

AAB2475922

 
alfa aesar™
B2475922
100g Each for $366.35
Description & Specifications

Specifications

Formula Weight 221.21
Chemical Name or Material N-(2-Methoxyphenyl)maleamic acid
Melting Point 143° to 145°C
Assay 97%
Quantity 5g
Molecular Formula C11H11NO4
MDL Number MFCD00020460
CAS 36847-94-6
Synonym 4-(2-Methoxyanilino)-4-oxobut-2-enoic acid
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.