missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-(2-Methoxyethyl)propylamine, TCI America™

Manufacturer:  TCI America M098025G

 View more versions of this product

Catalog No. M098025G



Specifications

Specifications

N-(2-Methoxyethyl)propylamine
Colorless
43175-57-1
MFCD00144830
UDZCEFCJEGGQOJ-UHFFFAOYSA-N
N-(2-methoxyethyl)propan-1-amine
12238049
Clear Liquid at 20°C
138°C
25g
C6H15NO
2733
CCCNCCOC
117.192
117.19
≥98.0% (GC,T)

Chemical Identifiers

43175-57-1
117.192
UDZCEFCJEGGQOJ-UHFFFAOYSA-N
N-(2-methoxyethyl)propan-1-amine
C6H15NO
MFCD00144830
12238049
CCCNCCOC
Safety and Handling

Safety and Handling

TSCA : No

Documents