missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-(2-Methoxyethyl)methylamine, TCI America™

Manufacturer:  TCI America M098625ML

 View more versions of this product

Catalog No. M098625ML



Specifications

Specifications

N-(2-Methoxyethyl)methylamine
Yellow
38256-93-8
MFCD00144829
KOHBEDRJXKOYHL-UHFFFAOYSA-N
2-methoxy-N-methylethanamine
300977
Clear Liquid at 20°C
98°C
25mL
C4H11NO
2924
CNCCOC
89.138
89.14
≥97.0% (GC,T)

Chemical Identifiers

38256-93-8
89.138
KOHBEDRJXKOYHL-UHFFFAOYSA-N
2-methoxy-N-methylethanamine
C4H11NO
MFCD00144829
300977
CNCCOC
Safety and Handling

Safety and Handling

TSCA : No

Documents