missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-(2-Methoxyethyl)ethylamine, TCI America™

Manufacturer:  TCI America E039725G

 View more versions of this product

Catalog No. E039725G



Specifications

Specifications

N-(2-Methoxyethyl)ethylamine
Yellow
34322-82-2
MFCD00144876
N-Ethyl-2-methoxyethylamine
CCNCCOC
103.165
103.17
≥97.0% (GC)
116°C
25g
C5H13NO
2734
VGEMYWDUTPQWBN-UHFFFAOYSA-N
N-ethyl-2-methoxyethanamine
546877
Clear Liquid at 20°C

Chemical Identifiers

34322-82-2
103.165
VGEMYWDUTPQWBN-UHFFFAOYSA-N
546877
CCNCCOC
C5H13NO
MFCD00144876
N-Ethyl-2-methoxyethylamine
N-ethyl-2-methoxyethanamine
Safety and Handling

Safety and Handling

TSCA : No

Documents