Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(2-Methoxy-5-methylphenyl)thiourea, 99%

Chemical Identifiers

Chemical Name or Material N-(2-Methoxy-5-methylphenyl)thiourea
CAS 88686-29-7
Molecular Formula C9H12N2OS
Formula Weight 196.27
MDL Number MFCD00041174
UN Number UN2811
Beilstein 2722947
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1210103

  SDS
alfa aesar™
L1210103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1210106

  SDS
alfa aesar™
L1210106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 196.27
Chemical Name or Material N-(2-Methoxy-5-methylphenyl)thiourea
CAS 88686-29-7
Assay 99%
Molecular Formula C9H12N2OS
UN Number UN2811
Beilstein 2722947
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301
Toxic if swallowed.
Health Hazard 3 GHS P Statement
P280f-P301+P310-P330-P501a
Wear protective clothing.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Melting Point 127° to 130°C
Quantity 1g
MDL Number MFCD00041174

Applications
N-(2-Methoxy-5-methylphenyl)thiourea is used as pharmaceutical intermediates.

Solubility
Insoluble in water.

Notes
Store in a cool, dry conditions in well sealed container. Incompatible with oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P280f-P301+P310-P330-P501a
Wear protective clothing.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301
Toxic if swallowed.