missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(2-Methoxy-5-methylphenyl)thiourea, 99%

Manufacturer:  Alfa Aesar™ L1210106

 View more versions of this product

Catalog No. AAL1210106



Description

Description

Applications
N-(2-Methoxy-5-methylphenyl)thiourea is used as pharmaceutical intermediates.

Solubility
Insoluble in water.

Notes
Store in a cool, dry conditions in well sealed container. Incompatible with oxidizing agents.
Specifications

Specifications

N-(2-Methoxy-5-methylphenyl)thiourea
5g
C9H12N2OS
UN2811
1-2-methoxy-5-methylphenyl-2-thiourea, 1-2-methoxy-5-methylphenyl thiourea, 2-methoxy-5-methylphenyl thiourea, amino 2-methoxy-5-methylphenyl amino methane-1-thione, acmc-20cw9w, 2-methoxy-5-methyl-phenyl-thiourea, n-2-methoxy-5-methylphenyl thiourea, 2-methoxy-5-methylphenylthiourea, thiourea,n-2-methoxy-5-methylphenyl
CC1=CC(=C(C=C1)OC)NC(=S)N
196.268
196.27
127°C to 130°C
88686-29-7
MFCD00041174
2722947
IPKUQBKZOAIUOC-UHFFFAOYSA-N
(2-methoxy-5-methylphenyl)thiourea
736212
99%

Chemical Identifiers

88686-29-7
196.268
IPKUQBKZOAIUOC-UHFFFAOYSA-N
736212
CC1=CC(=C(C=C1)OC)NC(=S)N
C9H12N2OS
MFCD00041174
1-2-methoxy-5-methylphenyl-2-thiourea, 1-2-methoxy-5-methylphenyl thiourea, 2-methoxy-5-methylphenyl thiourea, amino 2-methoxy-5-methylphenyl amino methane-1-thione, acmc-20cw9w, 2-methoxy-5-methyl-phenyl-thiourea, n-2-methoxy-5-methylphenyl thiourea, 2-methoxy-5-methylphenylthiourea, thiourea,n-2-methoxy-5-methylphenyl
(2-methoxy-5-methylphenyl)thiourea
Safety and Handling

Safety and Handling

GHS H Statement
H301
Toxic if swallowed.

GHS P Statement
P280f-P301+P310-P330-P501a
Wear protective clothing.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

Documents