missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(2-Methoxy-5-methylphenyl)benzenesulfonamide, 97%

$165.54 - $523.95

Chemical Identifiers

CAS 6964-02-9
Molecular Formula C8H18O2
Molecular Weight (g/mol) 146.23
MDL Number MFCD00085470
InChI Key ZMWAURRHTDCQCX-UHFFFAOYNA-N
Synonym 3-methyl-2,4-heptanediol
PubChem CID 248872
IUPAC Name 3-methylheptane-2,4-diol
SMILES CCCC(C(C)C(C)O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH55897MD View Documents Alfa Aesar™
H55897MD
250mg Each for $165.54
 
AAH5589703 View Documents Alfa Aesar™
H5589703
1g Each for $523.95
 
Specifications

Chemical Identifiers

6964-02-9
146.23
ZMWAURRHTDCQCX-UHFFFAOYNA-N
248872
CCCC(C(C)C(C)O)O
C8H18O2
MFCD00085470
3-methyl-2,4-heptanediol
3-methylheptane-2,4-diol

Specifications

250mg
C14H15NO3S
2'-methoxy-5'-methylbenzenesulfonanilide
CC1=CC(=C(C=C1)OC)NS(=O)(=O)C2=CC=CC=C2
277.34
277.34
N-(2-Methoxy-5-methylphenyl)benzenesulfonamide
6964-02-9
MFCD00085470
SNCSZUJMKFNFHX-UHFFFAOYSA-N
N-(2-methoxy-5-methylphenyl)benzenesulfonamide
248884
97%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures