missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(2-Methoxy-5-methylphenyl)benzenesulfonamide, 97%

Manufacturer:  Alfa Aesar™ H5589703

 View more versions of this product

Catalog No. AAH5589703



Specifications

Specifications

N-(2-Methoxy-5-methylphenyl)benzenesulfonamide
6964-02-9
MFCD00085470
ZMWAURRHTDCQCX-UHFFFAOYNA-N
3-methylheptane-2,4-diol
248872
97%
1g
C8H18O2
3-methyl-2,4-heptanediol
CCCC(C(C)C(C)O)O
146.23
277.34

Chemical Identifiers

6964-02-9
146.23
ZMWAURRHTDCQCX-UHFFFAOYNA-N
248872
CCCC(C(C)C(C)O)O
C8H18O2
MFCD00085470
3-methyl-2,4-heptanediol
3-methylheptane-2,4-diol
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents