missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-(2-Hydroxyethyl)pyrrolidine, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 121200250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
N-(2-Hydroxyethyl)pyrrolidine | |
94.0 | |
Brown to Yellow | |
79°C to 81°C (13.0 mmHg) | |
Authentic | |
Glass bottle | |
1.4713 to 1.4743 | |
25 g | |
0.98 | |
XBRDBODLCHKXHI-UHFFFAOYSA-N | |
2-pyrrolidin-1-ylethanol | |
76288 | |
115.17 | |
Liquid |
2955-88-6 | |
100.0 | |
0.9800g/mL | |
56°C | |
94% min. (GC) | |
C6H13NO | |
MFCD00003181 | |
20, II, 5 | |
1-pyrrolidineethanol, n-2-hydroxyethyl pyrrolidine, 1-2-hydroxyethyl pyrrolidine, 2-pyrrolidin-1-yl ethanol, epolamine, 2-pyrrolidinoethanol, pyrrolidinoethanol, hydroxyethylpyrrolidine, 2-1-pyrrolidinyl ethanol, 2-pyrrolidin-1-yl ethan-1-ol | |
OCCN1CCCC1 | |
115.18 | |
CHEBI:48293 | |
95% |
Chemical Identifiers
2955-88-6 | |
115.18 | |
XBRDBODLCHKXHI-UHFFFAOYSA-N | |
76288 | |
2-pyrrolidin-1-ylethanol |
C6H13NO | |
MFCD00003181 | |
1-pyrrolidineethanol, n-2-hydroxyethyl pyrrolidine, 1-2-hydroxyethyl pyrrolidine, 2-pyrrolidin-1-yl ethanol, epolamine, 2-pyrrolidinoethanol, pyrrolidinoethanol, hydroxyethylpyrrolidine, 2-1-pyrrolidinyl ethanol, 2-pyrrolidin-1-yl ethan-1-ol | |
CHEBI:48293 | |
OCCN1CCCC1 |
Safety and Handling
GHS H Statement
Flammable liquid and vapor.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Store in a well-ventilated place.
Keep container tightly closed.
GHS Signal Word: Warning
EINECSNumber : 220-976-3
RUO – Research Use Only