missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-(2-Hydroxyethyl)pyrrolidine, 95%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 121201000
Specifications
N-(2-Hydroxyethyl)pyrrolidine | |
56°C | |
Glass bottle | |
0.98 | |
Brown to Yellow | |
2955-88-6 | |
100.0 | |
C6H13NO | |
20, II, 5 | |
XBRDBODLCHKXHI-UHFFFAOYSA-N | |
2-pyrrolidin-1-ylethanol | |
76288 | |
115.17 | |
95% |
0.9800g/mL | |
Authentic | |
1.4713 to 1.4743 | |
79°C to 81°C (13.0mmHg) | |
100g | |
94.0 | |
94% min. (GC) | |
MFCD00003181 | |
1-2-hydroxyethyl pyrrolidine, 1-pyrrolidineethanol, 2-1-pyrrolidinyl ethanol, 2-pyrrolidin-1-yl ethan-1-ol, 2-pyrrolidin-1-yl ethanol, 2-pyrrolidinoethanol, epolamine, hydroxyethylpyrrolidine, n-2-hydroxyethyl pyrrolidine, pyrrolidinoethanol | |
C1CCN(C1)CCO | |
115.17 | |
CHEBI:48293 | |
Liquid |
Chemical Identifiers
2955-88-6 | |
115.17 | |
XBRDBODLCHKXHI-UHFFFAOYSA-N | |
76288 | |
2-pyrrolidin-1-ylethanol |
C6H13NO | |
MFCD00003181 | |
1-2-hydroxyethyl pyrrolidine, 1-pyrrolidineethanol, 2-1-pyrrolidinyl ethanol, 2-pyrrolidin-1-yl ethan-1-ol, 2-pyrrolidin-1-yl ethanol, 2-pyrrolidinoethanol, epolamine, hydroxyethylpyrrolidine, n-2-hydroxyethyl pyrrolidine, pyrrolidinoethanol | |
CHEBI:48293 | |
C1CCN(C1)CCO |
Safety and Handling
GHS H Statement
Flammable liquid and vapor.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Store in a well-ventilated place.
Keep container tightly closed.
GHS Signal Word: Warning
EINECSNumber : 220-976-3