missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-(2-Hydroxyethyl)phthalimide, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 124921000

 View more versions of this product

Catalog No. AC124921000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

N-(2-Hydroxyethyl)phthalimide
Glass bottle
126°C to 129°C
3891-07-4
100.0
C10H9NO3
+-n-pinacolylphthalamic acid, 2-1,2,2-trimethylpropyl amino carbonyl benzoic acid, 2-1,2,2-trimethylpropylcarbamoyl benzoic acid, 2-3,3-dimethylbutan-2-yl carbamoyl benzoic acid, 2-3,3-dimethylbutan-2-ylcarbamoyl benzoic acid, n-pinacolylphthalamic acid
OCCN1C(=O)C2=CC=CC=C2C1=O
191.19
191.19
99%
Authentic
White
100g
98.5
98.5% min. (on dry substance) (HPLC)
MFCD00005903
MWFLUYFYHANMCM-UHFFFAOYSA-N
2-(3,3-dimethylbutan-2-ylcarbamoyl)benzoic acid
3354762
Crystalline Powder

Chemical Identifiers

3891-07-4
191.19
MWFLUYFYHANMCM-UHFFFAOYSA-N
3354762
OCCN1C(=O)C2=CC=CC=C2C1=O
C10H9NO3
MFCD00005903
+-n-pinacolylphthalamic acid, 2-1,2,2-trimethylpropyl amino carbonyl benzoic acid, 2-1,2,2-trimethylpropylcarbamoyl benzoic acid, 2-3,3-dimethylbutan-2-yl carbamoyl benzoic acid, 2-3,3-dimethylbutan-2-ylcarbamoyl benzoic acid, n-pinacolylphthalamic acid
2-(3,3-dimethylbutan-2-ylcarbamoyl)benzoic acid
Safety and Handling

Safety and Handling

EINECSNumber : 223-434-4

SDS Documents