missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-(2-Hydroxyethyl)morpholine, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 121181000

 View more versions of this product

Catalog No. AC121181000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

N-(2-Hydroxyethyl)morpholine
107°C
Glass bottle
1.08
0.5% max.
Colorless to Yellow
100g
622-40-2
100.0
C6H13NO2
27, 7
Solubility in water: miscible
C1COCCN1CCO
131.17
CHEBI:67144
Liquid
1.0800g/mL
Authentic
1.4760 to 1.4780
26.6 mPa.s (20°C)
223.0°C to 225.0°C
-1.0°C
99%
98.5
99%
MFCD00006180
2-4-morpholinyl ethanol, 2-morpholinoethanol, 4-2-hydroxyethyl morpholine, 4-morpholineethanol, beta-morpholinoethanol, ethanol, 2-morpholino, hydroxyethylmorpholine, morpholine ethanol, morpholineethanol, n-2-hydroxyethyl morpholine
KKFDCBRMNNSAAW-UHFFFAOYSA-N
2-morpholin-4-ylethanol
61163
131.17
99%

Chemical Identifiers

622-40-2
131.17
KKFDCBRMNNSAAW-UHFFFAOYSA-N
61163
2-morpholin-4-ylethanol
C6H13NO2
MFCD00006180
2-4-morpholinyl ethanol, 2-morpholinoethanol, 4-2-hydroxyethyl morpholine, 4-morpholineethanol, beta-morpholinoethanol, ethanol, 2-morpholino, hydroxyethylmorpholine, morpholine ethanol, morpholineethanol, n-2-hydroxyethyl morpholine
CHEBI:67144
C1COCCN1CCO
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 210-734-5

RTECSNumber : QE3510000

TSCA : TSCA

SDS Documents
Product Certifications