missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-(2-Hydroxyethyl)maleimide, TCI America™

$278.06 - $832.66

Chemical Identifiers

CAS 1585-90-6
Molecular Formula C6H7NO3
Molecular Weight (g/mol) 141.126
MDL Number MFCD00465266
InChI Key AXTADRUCVAUCRS-UHFFFAOYSA-N
Synonym HEMI
PubChem CID 458487
IUPAC Name 1-(2-hydroxyethyl)pyrrole-2,5-dione
SMILES C1=CC(=O)N(C1=O)CCO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H15441G View Documents TCI America
H15441G
1g Each for $278.06
 
H15445G View Documents TCI America
H15445G
5g Each for $832.66
 
Specifications

Chemical Identifiers

1585-90-6
141.126
AXTADRUCVAUCRS-UHFFFAOYSA-N
458487
C1=CC(=O)N(C1=O)CCO
C6H7NO3
MFCD00465266
HEMI
1-(2-hydroxyethyl)pyrrole-2,5-dione

Specifications

130°C
73°C
1585-90-6
MFCD00465266
AXTADRUCVAUCRS-UHFFFAOYSA-N
1-(2-hydroxyethyl)pyrrole-2,5-dione
458487
Crystal-Powder at 20°C
N-(2-Hydroxyethyl)maleimide
White-Yellow
1g
C6H7NO3
HEMI
C1=CC(=O)N(C1=O)CCO
141.126
141.13
≥98.0% (HPLC,N)
Safety and Handling

Safety and Handling

TSCA : No