Learn More
N-(2-Hydroxyethyl)hexamethyleneimine, 95%, Thermo Scientific Chemicals
$64.96 - $207.99
Chemical Identifiers
CAS | 20603-00-3 |
---|---|
Molecular Formula | C8H17NO |
Molecular Weight (g/mol) | 143.23 |
MDL Number | MFCD00020988 |
InChI Key | VMRYMOMQCYSPHS-UHFFFAOYSA-N |
Synonym | 2-azepan-1-yl ethanol, hexahydro-1h-azepine-1-ethanol, n-2-hydroxyethyl hexamethyleneimine, 2-azepan-1-yl ethan-1-ol, 2-hexamethyleneimino ethanol, 2-azaperhydroepinylethan-1-ol, 2-azepan-1-ylethan-1-ol, 2-azepan-1-yl ;ethanol, 2-n-hexamethyleneimino ethanol, 1-2-hydroxyethyl homopiperidine |
PubChem CID | 88615 |
IUPAC Name | 2-(azepan-1-yl)ethanol |
SMILES | C1CCCN(CC1)CCO |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL0649706
|
Thermo Scientific Chemicals
L0649706 |
5 g |
Each for $64.96
|
|
|||||
AAL0649714
|
Thermo Scientific Chemicals
L0649714 |
25 g |
Each for $207.99
|
|
|||||
Description
N-(2-Hydroxyethyl)hexamethyleneimine is used in organic synthesis, building blocks. It is mainly involved as an intermediate in the synthesis of Bazedoxifene acetate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsN-(2-Hydroxyethyl)hexamethyleneimine is used in organic synthesis, building blocks. It is mainly involved as an intermediate in the synthesis of Bazedoxifene acetate.
Solubility
Fully miscible in water.
Notes
Store away from air and oxidizing agents. Store under dry inert gas. Incompatible with air and oxidizing agents.
Chemical Identifiers
20603-00-3 | |
143.23 | |
VMRYMOMQCYSPHS-UHFFFAOYSA-N | |
88615 | |
C1CCCN(CC1)CCO |
C8H17NO | |
MFCD00020988 | |
2-azepan-1-yl ethanol, hexahydro-1h-azepine-1-ethanol, n-2-hydroxyethyl hexamethyleneimine, 2-azepan-1-yl ethan-1-ol, 2-hexamethyleneimino ethanol, 2-azaperhydroepinylethan-1-ol, 2-azepan-1-ylethan-1-ol, 2-azepan-1-yl ;ethanol, 2-n-hexamethyleneimino ethanol, 1-2-hydroxyethyl homopiperidine | |
2-(azepan-1-yl)ethanol |
Specifications
20603-00-3 | |
114°C to 115°C (23 mmHg) | |
1.484 | |
5 g | |
104110 | |
2-azepan-1-yl ethanol, hexahydro-1h-azepine-1-ethanol, n-2-hydroxyethyl hexamethyleneimine, 2-azepan-1-yl ethan-1-ol, 2-hexamethyleneimino ethanol, 2-azaperhydroepinylethan-1-ol, 2-azepan-1-ylethan-1-ol, 2-azepan-1-yl ;ethanol, 2-n-hexamethyleneimino ethanol, 1-2-hydroxyethyl homopiperidine | |
VMRYMOMQCYSPHS-UHFFFAOYSA-N | |
2-(azepan-1-yl)ethanol | |
88615 | |
95% |
0.971 | |
C8H17NO | |
MFCD00020988 | |
UN2735 | |
Air Sensitive | |
Fully miscible in water. | |
C1CCCN(CC1)CCO | |
143.23 | |
143.22 | |
N-(2-Hydroxyethyl)hexamethyleneimine |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302+H312-H314
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
EINECSNumber : 243-915-2
TSCA : No
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only