missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-(2-Hydroxyethyl)acrylamide (stabilized with MEHQ), TCI America™

$38.19 - $157.36

Chemical Identifiers

CAS 7646-67-5
Molecular Formula C5H9NO2
Molecular Weight (g/mol) 115.132
InChI Key UUORTJUPDJJXST-UHFFFAOYSA-N
PubChem CID 193628
IUPAC Name N-(2-hydroxyethyl)prop-2-enamide
SMILES C=CC(=O)NCCO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H126225G View Documents TCI America
H126225G
25g Each for $38.19
 
H1262500G View Documents TCI America
H1262500G
500g Each for $157.36
 
Specifications

Chemical Identifiers

7646-67-5
115.132
193628
C=CC(=O)NCCO
C5H9NO2
UUORTJUPDJJXST-UHFFFAOYSA-N
N-(2-hydroxyethyl)prop-2-enamide

Specifications

130°C
25g
C5H9NO2
C=CC(=O)NCCO
115.132
115.13
≥98.0% (GC,T)
Undesignated
7646-67-5
UUORTJUPDJJXST-UHFFFAOYSA-N
N-(2-hydroxyethyl)prop-2-enamide
193628
Clear Liquid at 20°C
N-(2-Hydroxyethyl)acrylamide (stabilized with MEHQ)
Safety and Handling

Safety and Handling

EINECSNumber : (2)-1023

TSCA : No

Recommended Storage : Refrigerator