missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(2-Hydroxybenzoyl)pyrrolidine, 97%

Manufacturer:  Alfa Aesar™ H5569506

 View more versions of this product

Catalog No. AAH5569506



Specifications

Specifications

N-(2-Hydroxybenzoyl)pyrrolidine
118°C to 124°C
98841-68-0
MFCD00020832
LELBEJMWNJWJDG-UHFFFAOYSA-N
(2-hydroxyphenyl)-pyrrolidin-1-ylmethanone
225361
Solid
Brown to White
5g
C11H13NO2
1-2-hydroxybenzoyl pyrrolidine
C1CCN(C1)C(=O)C2=CC=CC=C2O
191.23
191.23
97%

Chemical Identifiers

98841-68-0
191.23
LELBEJMWNJWJDG-UHFFFAOYSA-N
225361
C1CCN(C1)C(=O)C2=CC=CC=C2O
C11H13NO2
MFCD00020832
1-2-hydroxybenzoyl pyrrolidine
(2-hydroxyphenyl)-pyrrolidin-1-ylmethanone
Safety and Handling

Safety and Handling

GHS H Statement
H302-H319
Harmful if swallowed.
Causes serious eye irritation.

GHS P Statement
P280-P264-P305+P351+P338-P301+P312-P337+P313-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
If eye irritation persists: Get medical advice/attention.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents