missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(2-Hydroxybenzoyl)pyrrolidine, 97%

$46.93 - $176.80

Chemical Identifiers

CAS 98841-68-0
Molecular Formula C11H13NO2
Molecular Weight (g/mol) 191.23
MDL Number MFCD00020832
InChI Key LELBEJMWNJWJDG-UHFFFAOYSA-N
Synonym 1-2-hydroxybenzoyl pyrrolidine
PubChem CID 225361
IUPAC Name (2-hydroxyphenyl)-pyrrolidin-1-ylmethanone
SMILES C1CCN(C1)C(=O)C2=CC=CC=C2O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5569506 View Documents Alfa Aesar™
H5569506
5g Each for $46.93
 
AAH5569514 View Documents Alfa Aesar™
H5569514
25g Each for $176.80
 
Specifications

Chemical Identifiers

98841-68-0
191.23
LELBEJMWNJWJDG-UHFFFAOYSA-N
225361
C1CCN(C1)C(=O)C2=CC=CC=C2O
C11H13NO2
MFCD00020832
1-2-hydroxybenzoyl pyrrolidine
(2-hydroxyphenyl)-pyrrolidin-1-ylmethanone

Specifications

Brown to White
5g
C11H13NO2
1-2-hydroxybenzoyl pyrrolidine
C1CCN(C1)C(=O)C2=CC=CC=C2O
191.23
191.23
97%
118°C to 124°C
98841-68-0
MFCD00020832
LELBEJMWNJWJDG-UHFFFAOYSA-N
(2-hydroxyphenyl)-pyrrolidin-1-ylmethanone
225361
Solid
N-(2-Hydroxybenzoyl)pyrrolidine
Safety and Handling

Safety and Handling

GHS H Statement
H302-H319
Harmful if swallowed.
Causes serious eye irritation.

P264b-P270-P280i-P301+P312-P305+P351+P338-P330-P501c

H302-H319

TSCA : No

Recommended Storage : Ambient temperatures