missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-(2-Hydroxy-3-sulfopropyl)-3,5-dimethoxyaniline Sodium Salt [for Biochemical Research], TCI America™

Manufacturer:  TCI America S08271G

 View more versions of this product

Catalog No. S08271G



Specifications

Specifications

N-(2-Hydroxy-3-sulfopropyl)-3,5-dimethoxyaniline Sodium Salt [for Biochemical Research]
1g
C11H16NNaO6S
3-(3,5-Dimethoxyanilino)-2-hydroxypropanesulfonic Acid Sodium Salt, Sodium 3-(3,5-Dimethoxyanilino)-2-hydroxypropanesulfonate, HDAOS
COC1=CC(=CC(=C1)NCC(CS(=O)(=O)[O-])O)OC.[Na+]
313.3
313.30
≥98.0% (HPLC)
White
82692-88-4
MFCD00063425
SVLRFMQGKVFRTB-UHFFFAOYSA-M
sodium;3-(3,5-dimethoxyanilino)-2-hydroxypropane-1-sulfonate
23666667
Crystal-Powder at 20°C

Chemical Identifiers

82692-88-4
313.3
SVLRFMQGKVFRTB-UHFFFAOYSA-M
23666667
COC1=CC(=CC(=C1)NCC(CS(=O)(=O)[O-])O)OC.[Na+]
C11H16NNaO6S
MFCD00063425
3-(3,5-Dimethoxyanilino)-2-hydroxypropanesulfonic Acid Sodium Salt, Sodium 3-(3,5-Dimethoxyanilino)-2-hydroxypropanesulfonate, HDAOS
sodium;3-(3,5-dimethoxyanilino)-2-hydroxypropane-1-sulfonate
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

Documents