Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(2-Fluorophenyl)thiourea, 97%

Chemical Identifiers

Chemical Name or Material N-(2-Fluorophenyl)thiourea
CAS 656-32-6
Molecular Formula C7H7FN2S
Formula Weight 170.21
MDL Number MFCD00041148
UN Number UN2811
Beilstein 2718420
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2088303

  SDS
alfa aesar™
B2088303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2088306

  SDS
alfa aesar™
B2088306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 170.21
Chemical Name or Material N-(2-Fluorophenyl)thiourea
Assay 97%
Health Hazard 1 Danger
Odor Odorless
Quantity 1g
Beilstein 2718420
Melting Point 141° to 148°C
Health Hazard 2 GHS H Statement
H301
Toxic if swallowed.
UN Number UN2811
Molecular Formula C7H7FN2S
MDL Number MFCD00041148
Health Hazard 3 GHS P Statement
P280f-P309-P310
Wear protective clothing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
CAS 656-32-6
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P280f-P309-P310
Wear protective clothing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H301
Toxic if swallowed.