missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(2-Ethoxyphenyl)-3-methylbenzamide, 97%

Chemical Identifiers

CAS 349096-36-2
Molecular Formula C16H17NO2
Molecular Weight (g/mol) 255.317
MDL Number MFCD00784144
InChI Key WVKNKVSKPRDADI-UHFFFAOYSA-N
Synonym n-2-ethoxyphenyl-3-methylbenzamide, cbmicro_018461
PubChem CID 2842554
IUPAC Name N-(2-ethoxyphenyl)-3-methylbenzamide
SMILES CCOC1=CC=CC=C1NC(=O)C2=CC(=CC=C2)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH59872MD View Documents Alfa Aesar™
H59872MD
250mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAH5987203 View Documents Alfa Aesar™
H5987203
1g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

349096-36-2
255.317
WVKNKVSKPRDADI-UHFFFAOYSA-N
2842554
CCOC1=CC=CC=C1NC(=O)C2=CC(=CC=C2)C
C16H17NO2
MFCD00784144
n-2-ethoxyphenyl-3-methylbenzamide, cbmicro_018461
N-(2-ethoxyphenyl)-3-methylbenzamide

Specifications

250mg
C16H17NO2
n-2-ethoxyphenyl-3-methylbenzamide, cbmicro_018461
CCOC1=CC=CC=C1NC(=O)C2=CC(=CC=C2)C
255.317
255.32
N-(2-Ethoxyphenyl)-3-methylbenzamide
349096-36-2
MFCD00784144
WVKNKVSKPRDADI-UHFFFAOYSA-N
N-(2-ethoxyphenyl)-3-methylbenzamide
2842554
97%