missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(2-Ethoxyphenyl)-2-methoxybenzamide, 97%

$211.27 - $675.45

Chemical Identifiers

CAS 306279-65-2
Molecular Formula C16H17NO3
Molecular Weight (g/mol) 271.316
MDL Number MFCD01010708
InChI Key IFOHCGGZEGVBME-UHFFFAOYSA-N
Synonym n-2-ethoxyphenyl-2-methoxybenzamide, n-2-ethoxyphenyl-2-methoxybenzamide, n-2-ethoxyphenyl 2-methoxyphenyl carboxamide
PubChem CID 700574
IUPAC Name N-(2-ethoxyphenyl)-2-methoxybenzamide
SMILES CCOC1=CC=CC=C1NC(=O)C2=CC=CC=C2OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH59799MD View Documents Alfa Aesar™
H59799MD
250mg Each for $211.27
 
AAH5979903 View Documents Alfa Aesar™
H5979903
1g Each for $675.45
 
Specifications

Chemical Identifiers

306279-65-2
271.316
IFOHCGGZEGVBME-UHFFFAOYSA-N
700574
CCOC1=CC=CC=C1NC(=O)C2=CC=CC=C2OC
C16H17NO3
MFCD01010708
n-2-ethoxyphenyl-2-methoxybenzamide, n-2-ethoxyphenyl-2-methoxybenzamide, n-2-ethoxyphenyl 2-methoxyphenyl carboxamide
N-(2-ethoxyphenyl)-2-methoxybenzamide

Specifications

250mg
C16H17NO3
n-2-ethoxyphenyl-2-methoxybenzamide, n-2-ethoxyphenyl-2-methoxybenzamide, n-2-ethoxyphenyl 2-methoxyphenyl carboxamide
CCOC1=CC=CC=C1NC(=O)C2=CC=CC=C2OC
271.316
271.32
N-(2-Ethoxyphenyl)-2-methoxybenzamide
306279-65-2
MFCD01010708
IFOHCGGZEGVBME-UHFFFAOYSA-N
N-(2-ethoxyphenyl)-2-methoxybenzamide
700574
97%