Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(2-Cyanoethyl)-N-methylaniline, 97%

$34.32 - $90.50

Chemical Identifiers

Chemical Name or Material N-(2-Cyanoethyl)-N-methylaniline
CAS 94-34-8
Molecular Formula C10H12N2
Formula Weight 160.22
MDL Number MFCD00019856
Beilstein 2803416
Synonym 3-(N-Methylanilino)propionitrile
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1116206

  SDS
alfa aesar™
L1116206
5g Each for $34.32

AAL1116214

  SDS
alfa aesar™
L1116214
25g Each for $90.50
Description & Specifications

Specifications

Formula Weight 160.22
Chemical Name or Material N-(2-Cyanoethyl)-N-methylaniline
Assay 97%
MDL Number MFCD00019856
CAS 94-34-8
Synonym 3-(N-Methylanilino)propionitrile
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.
Health Hazard 3 GHS P Statement
P280h
Wear protective gloves/clothing.
TSCA TSCA
Molecular Formula C10H12N2
Boiling Point 125° to 127°C (4mmHg)
Beilstein 2803416
Quantity 5g
Flash Point >110°C (230°F)
Safety and Handling

Warning

GHS P Statement
P280h
Wear protective gloves/clothing.

GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.