missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(2-Cyanoethyl)-N-methylaniline, 97%

Manufacturer:  Alfa Aesar™ L1116206

 View more versions of this product

Catalog No. AAL1116206



Specifications

Specifications

N-(2-Cyanoethyl)-N-methylaniline
125°C to 127°C (4mmHg)
94-34-8
MFCD00019856
n-2-cyanoethyl-n-methylaniline, 3-methyl phenyl amino propanenitrile, propanenitrile, 3-methylphenylamino, n-methyl-n-2-cyanoethyl aniline, n-cyanoethyl-n-methylaniline, 3-n-methyl-n-phenylamino propionitrile, 3-methyl phenyl amino propanenitrile, 3-n-methylanilino propionitrile, 3-n-methylanilino propanenitrile, n-.beta.-cyanoethyl-n-methylaniline
CN(CCC#N)C1=CC=CC=C1
160.22
160.22
>110°C (230°F)
5g
C10H12N2
2803416
IXXLKTZOCSRXEM-UHFFFAOYSA-N
3-(N-methylanilino)propanenitrile
66748
97%

Chemical Identifiers

94-34-8
160.22
IXXLKTZOCSRXEM-UHFFFAOYSA-N
66748
CN(CCC#N)C1=CC=CC=C1
C10H12N2
MFCD00019856
n-2-cyanoethyl-n-methylaniline, 3-methyl phenyl amino propanenitrile, propanenitrile, 3-methylphenylamino, n-methyl-n-2-cyanoethyl aniline, n-cyanoethyl-n-methylaniline, 3-n-methyl-n-phenylamino propionitrile, 3-methyl phenyl amino propanenitrile, 3-n-methylanilino propionitrile, 3-n-methylanilino propanenitrile, n-.beta.-cyanoethyl-n-methylaniline
3-(N-methylanilino)propanenitrile
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
P280h
Wear protective gloves/clothing.

Warning

TSCA : TSCA

Documents