Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(2-Cyanoethyl)-N-(2-hydroxyethyl)aniline, 98%

$106.68 - $106.68

Chemical Identifiers

Chemical Name or Material N-(2-Cyanoethyl)-N-(2-hydroxyethyl)aniline
CAS 92-64-8
Molecular Formula C11H14N2O
Formula Weight 190.25
MDL Number MFCD00035705
Beilstein 2369849
Synonym 3-[N-(2-Hydoxyethyl)anilino]propionitrile
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1118318

  SDS
alfa aesar™
L1118318
50g Each for $106.68
Description & Specifications

Specifications

Formula Weight 190.25
Chemical Name or Material N-(2-Cyanoethyl)-N-(2-hydroxyethyl)aniline
Molecular Formula C11H14N2O
CAS 92-64-8
TSCA TSCA
MDL Number MFCD00035705
Boiling Point 107° to 110°C (15mmHg)
Beilstein 2369849
Quantity 50g
Synonym 3-[N-(2-Hydoxyethyl)anilino]propionitrile
RTECS Number UG2905000
Assay 98%

Applications
N-(2-Cyanoethyl)-N-(2-hydroxyethyl)aniline is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Not miscible or difficult to mix with water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with bases and oxidizing agents.