missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-(2-Cyanoethyl)-N-(2-hydroxyethyl)aniline, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals L1118318

 View more versions of this product

Catalog No. AAL1118318



Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
N-(2-Cyanoethyl)-N-(2-hydroxyethyl)aniline is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Not miscible or difficult to mix with water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with bases and oxidizing agents.
Specifications

Specifications

N-(2-Cyanoethyl)-N-(2-hydroxyethyl)aniline
1.12
C11H14N2O
MFCD00035705
2369849
Not miscible or difficult to mix with water.
C1=CC=C(C=C1)N(CCC#N)CCO
190.246
190.25
92-64-8
107°C to 110°C (15 mmHg)
1.5705
50 g
n-cyanoethyl-n-hydroxyethylaniline, n-2-cyanoethyl-n-2-hydroxyethyl aniline, emery 5724, n-cyanoethyl-hydroxyethyl aniline, propanenitrile, 3-2-hydroxyethyl phenylamino, n-cyanoethyl-n-hydroxyethyl aniline, aniline, n-cyanoethyl-n-hydroxyethyl, 3-2-hydroxyethyl phenylamino propanenitrile, 3-n-2-hydroxyethyl anilino propiononitrile, propionitrile, 3-2-hydroxyethyl phenylamino
WZJJWQVBLSPALW-UHFFFAOYSA-N
3-[N-(2-hydroxyethyl)anilino]propanenitrile
113349
98%

Chemical Identifiers

92-64-8
190.246
WZJJWQVBLSPALW-UHFFFAOYSA-N
113349
C1=CC=C(C=C1)N(CCC#N)CCO
C11H14N2O
MFCD00035705
n-cyanoethyl-n-hydroxyethylaniline, n-2-cyanoethyl-n-2-hydroxyethyl aniline, emery 5724, n-cyanoethyl-hydroxyethyl aniline, propanenitrile, 3-2-hydroxyethyl phenylamino, n-cyanoethyl-n-hydroxyethyl aniline, aniline, n-cyanoethyl-n-hydroxyethyl, 3-2-hydroxyethyl phenylamino propanenitrile, 3-n-2-hydroxyethyl anilino propiononitrile, propionitrile, 3-2-hydroxyethyl phenylamino
3-[N-(2-hydroxyethyl)anilino]propanenitrile
Safety and Handling

Safety and Handling

EINECSNumber : 202-174-5

RTECSNumber : UG2905000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions