missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(2-Cyanoethyl)-N-(2-hydroxyethyl)aniline, 98%

Manufacturer:  Alfa Aesar™ L1118309

 View more versions of this product

Catalog No. AAL1118309



Description

Description

Applications
N-(2-Cyanoethyl)-N-(2-hydroxyethyl)aniline is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Not miscible or difficult to mix with water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with bases and oxidizing agents.
Specifications

Specifications

N-(2-Cyanoethyl)-N-(2-hydroxyethyl)aniline
10g
C11H14N2O
2369849
WZJJWQVBLSPALW-UHFFFAOYSA-N
3-[N-(2-hydroxyethyl)anilino]propanenitrile
113349
98%
107°C to 110°C (15mmHg)
92-64-8
MFCD00035705
n-cyanoethyl-n-hydroxyethylaniline, n-2-cyanoethyl-n-2-hydroxyethyl aniline, emery 5724, n-cyanoethyl-hydroxyethyl aniline, propanenitrile, 3-2-hydroxyethyl phenylamino, n-cyanoethyl-n-hydroxyethyl aniline, aniline, n-cyanoethyl-n-hydroxyethyl, 3-2-hydroxyethyl phenylamino propanenitrile, 3-n-2-hydroxyethyl anilino propiononitrile, propionitrile, 3-2-hydroxyethyl phenylamino
C1=CC=C(C=C1)N(CCC#N)CCO
190.246
190.25

Chemical Identifiers

92-64-8
190.246
WZJJWQVBLSPALW-UHFFFAOYSA-N
113349
C1=CC=C(C=C1)N(CCC#N)CCO
C11H14N2O
MFCD00035705
n-cyanoethyl-n-hydroxyethylaniline, n-2-cyanoethyl-n-2-hydroxyethyl aniline, emery 5724, n-cyanoethyl-hydroxyethyl aniline, propanenitrile, 3-2-hydroxyethyl phenylamino, n-cyanoethyl-n-hydroxyethyl aniline, aniline, n-cyanoethyl-n-hydroxyethyl, 3-2-hydroxyethyl phenylamino propanenitrile, 3-n-2-hydroxyethyl anilino propiononitrile, propionitrile, 3-2-hydroxyethyl phenylamino
3-[N-(2-hydroxyethyl)anilino]propanenitrile
Safety and Handling

Safety and Handling

RTECSNumber : UG2905000

TSCA : TSCA

Documents