missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-(2-Cyanoethyl)glycine, 98%, Acros Organics

Manufacturer:  Thermo Fisher Scientific AC154901000

 View more versions of this product

Catalog No. AC154901000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

N-(2-Cyanoethyl)glycine
Glass bottle
188°C to 193°C
98%
97.5
98%
NCCH2CH2NHCH2CO2H
.alpha.-.beta.'-cyanoethyl-aminoacetic acid
C(CNCC(=O)O)C#N
128.13
128.13
98%
Authentic
White
100g
3088-42-4
100.0
C5H8N2O2
MFCD00066903
KZUBZCHAWPDYQX-UHFFFAOYSA-N
2-(2-cyanoethylamino)acetic acid
76526
Powder or Flakes

Chemical Identifiers

3088-42-4
128.13
KZUBZCHAWPDYQX-UHFFFAOYSA-N
76526
C(CNCC(=O)O)C#N
C5H8N2O2
MFCD00066903
.alpha.-.beta.'-cyanoethyl-aminoacetic acid
2-(2-cyanoethylamino)acetic acid
Safety and Handling

Safety and Handling

GHS H Statement
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Avoid breath

GHS Signal Word: Warning

EINECSNumber : 221-418-1

SDS Documents