missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-(2-Cyanoethyl)glycine, 98%, Thermo Scientific™
Chemical Identifiers
CAS | 3088-42-4 |
---|---|
Molecular Formula | C5H8N2O2 |
Molecular Weight (g/mol) | 128.13 |
MDL Number | MFCD00066903 |
InChI Key | KZUBZCHAWPDYQX-UHFFFAOYSA-N |
Synonym | n-2-cyanoethyl glycine, glycine, n-2-cyanoethyl, n-2-cyanoethyl aminoacetic acid, 2-2-cyanoethyl amino acetic acid, 2-2-cyanoethylamino acetic acid, 2-cyanoethyl amino acetic acid, wln: qv1m2cn, .alpha.-.beta.'-cyanoethyl-aminoacetic acid |
PubChem CID | 76526 |
IUPAC Name | 2-(2-cyanoethylamino)acetic acid |
SMILES | OC(=O)CNCCC#N |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC154901000
|
Thermo Scientific Chemicals
154901000 |
100g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
3088-42-4 | |
128.13 | |
KZUBZCHAWPDYQX-UHFFFAOYSA-N | |
76526 | |
OC(=O)CNCCC#N |
C5H8N2O2 | |
MFCD00066903 | |
n-2-cyanoethyl glycine, glycine, n-2-cyanoethyl, n-2-cyanoethyl aminoacetic acid, 2-2-cyanoethyl amino acetic acid, 2-2-cyanoethylamino acetic acid, 2-cyanoethyl amino acetic acid, wln: qv1m2cn, .alpha.-.beta.'-cyanoethyl-aminoacetic acid | |
2-(2-cyanoethylamino)acetic acid |
Specifications
3088-42-4 | |
100.0 | |
98% | |
C5H8N2O2 | |
MFCD00066903 | |
n-2-cyanoethyl glycine, glycine, n-2-cyanoethyl, n-2-cyanoethyl aminoacetic acid, 2-2-cyanoethyl amino acetic acid, 2-2-cyanoethylamino acetic acid, 2-cyanoethyl amino acetic acid, wln: qv1m2cn, .alpha.-.beta.'-cyanoethyl-aminoacetic acid | |
OC(=O)CNCCC#N | |
128.13 | |
128.13 | |
N-(2-Cyanoethyl)glycine, 98% |
97.5 | |
Authentic | |
Glass bottle | |
NCCH2CH2NHCH2CO2H | |
100g | |
KZUBZCHAWPDYQX-UHFFFAOYSA-N | |
2-(2-cyanoethylamino)acetic acid | |
76526 | |
98% |
Safety and Handling
GHS H Statement
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Avoid breath
GHS Signal Word: Warning
EINECSNumber : 221-418-1
RUO â Research Use Only